Invitrogen
Manufacturer Code:PA5100467
Catalog # PIPA5100467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DDRGK1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110001I20Rik 2600009E05Rik AI326138 C20orf116 Dashurin DDRGK domain containing 1 DDRGK domain-containing protein 1 Ddrgk1 dJ1187M17.3 RGD1309979 Ufbp1 UFM1-binding and PCI domain-containing protein 1 UFM1-binding protein 1 containing a PCI domain; Dashurin; DDRGK domain-containing protein 1; UFM1-binding and PCI domain-containing protein 1; UFM1-binding protein 1 containing a PCI domain
Gene Aliases: 1110001I20Rik; 2600009E05Rik; AI326138; C20orf116; DDRGK1; dJ1187M17.3; UFBP1
UniProt ID: (Human) Q96HY6, (Mouse) Q80WW9
Entrez Gene ID: (Human) 65992, (Mouse) 77006
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.