Novus Biologicals
Manufacturer Code:NBP15937820UL
Catalog # NBP15937820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to C20ORF116 The peptide sequence was selected from the middle region of C20ORF116. Peptide sequence EERKRLESQREAEWKKEEERLRLEEEQKEEEERKAREEQAQREHEEYLKL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf116 chromosome 20 open reading frame 116 DDRGK domain containing 1 DDRGK domain-containing protein 1 dJ1187M17.3 MGC2592; Dashurin; DDRGK domain-containing protein 1; UFM1-binding and PCI domain-containing protein 1; UFM1-binding protein 1 containing a PCI domain
Gene Aliases: C20orf116; DDRGK1; dJ1187M17.3; UFBP1
UniProt ID: (Human) Q96HY6
Entrez Gene ID: (Human) 65992
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.