Invitrogen
Manufacturer Code:MA515356
Catalog # PIMA515356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B11E4 |
Immunogen | Purified recombit fragment of human DDR2 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD167 antigen-like family member B; CD167b; cell migration-inducing protein 20; Discoidin domain receptor 2; discoidin domain receptor family, member 2; Discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein kinase; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; Neurotrophic tyrosine kinase, receptor-related 3; Receptor protein-tyrosine kinase TKT; TKT MIG20a NTRKR3 TYRO10 DDR2; Tyrosine-protein kinase TYRO10; tyrosylprotein kinase
Gene Aliases: DDR2; MIG20a; NTRKR3; TKT; TYRO10
UniProt ID: (Human) Q16832
Entrez Gene ID: (Human) 4921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.