R&D Systems
Manufacturer Code:MAB25382
Catalog # MAB25382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1119D |
Immunogen | Phosphopeptide containing the human DDR2 Y740 site Accession # Q16832 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after opening. 6 months -20 to -70 degreesC under sterile conditions |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD167 antigen-like family member B; CD167 antigen-like family member B CD167b antigen Discoidin domain receptor 2 discoidin domain receptor family member 2 discoidin domain receptor tyrosine kinase 2 discoidin domain-containing receptor 2 EC 2.7.10 EC 2.7.10.1 hydroxyaryl-protein kinase migration-inducing gene 16 protein neurotrophic tyrosine kinase receptor related 3 Neurotrophic tyrosine kinase receptor-related 3 NTRKR3cell migration-inducing protein 20 Receptor protein-tyrosine kinase TKT TKT TKTMIG20a Trk3 Tyro-10 TYRO10 Tyrosine-protein kinase TYRO10 tyrosylprotein kinase; CD167b; cell migration-inducing protein 20; Discoidin domain receptor 2; discoidin domain receptor family, member 2; Discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein kinase; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; Neurotrophic tyrosine kinase, receptor-related 3; Receptor protein-tyrosine kinase TKT; Tyrosine-protein kinase TYRO10; tyrosylprotein kinase
Gene Aliases: DDR2; MIG20a; NTRKR3; TKT; TYRO10
UniProt ID: (Human) Q16832
Entrez Gene ID: (Human) 4921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.