Invitrogen
Manufacturer Code:PA527752
Catalog # PIPA527752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 628 and 855 of Human DDR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD167 antigen-like family member B; CD167b; cell migration-inducing protein 20; Discoidin domain receptor 2; discoidin domain receptor family, member 2; Discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein kinase; MIG20a NTRKR3 TKT TYRO10 CD167 antigen-like family member B cell migration-inducing protein 20 discoidin domain receptor 2 discoidin domain receptor family member 2 discoidin domain-containing receptor 2 discoidin domain-containing receptor tyrosine kinase 2 hydroxyaryl-protein; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; Neurotrophic tyrosine kinase, receptor-related 3; Receptor protein-tyrosine kinase TKT; Tyrosine-protein kinase TYRO10; tyrosylprotein kinase
Gene Aliases: AW495251; DDR2; MIG20a; NTRKR3; TKT; TYRO10
UniProt ID: (Human) Q16832, (Mouse) Q62371
Entrez Gene ID: (Human) 4921, (Mouse) 18214
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.