Invitrogen
Manufacturer Code:PA542898
Catalog # PIPA542898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DDOST |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: advanced glycation end-product receptor 1; advanced glycation endproduct receptor 1; DDOST 48 kDa subunit; dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide protein glycosyltransferase; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase AGE-R1 KIAA0115 MGC2191 OK/SW-cl.45 OST OST48 WBP1 AGER1 OKSWcl45; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48
Gene Aliases: AGER1; CDG1R; DDOST; KIAA0115; OK/SW-cl.45; OKSWcl45; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:77478
UniProt ID: (Dog) Q05052, (Human) P39656, (Bovine) A6QPY0, (Mouse) O54734, (Rat) Q641Y0, (Zebrafish) Q6NYS8
Entrez Gene ID: (Sheep) 100037660, (Horse) 100071776, (Rabbit) 100341880, (Dog) 404012, (Human) 1650, (Bovine) 510682, (Guinea pig) 100719976, (Mouse) 13200, (Rat) 313648, (Zebrafish) 406408
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.