Novus Biologicals
Manufacturer Code:NBP169562
Catalog # NBP169562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DDOST(dolichyl-diphosphooligosaccharide-protein glycosyltransferase) The peptide sequence was selected from the N terminal of DDOST. Peptide sequence SPSVEDFGGNINVETISAFIDGGGSVLVAASSDIGDPLRELGSECGIEFD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: advanced glycation end-product receptor 1; advanced glycation endproduct receptor 1; advanced glycation endproduct receptor 1 DDOST 48 kDa subunit dolichyl-diphosphooligosaccharide-protein glycosyltransferase dolichyl-diphosphooligosaccharide--protein glycosyltransferase dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit dolichyl-diphosphooligosaccharide-protein glycotransferase EC 2.4.1.119 KIAA0115MGC2191 Oligosaccharyl transferase 48 kDa subunit oligosaccharyltransferase 48 kDa subunit OST48AGER1 OSTOKSWcl45 WBP1; DDOST 48 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-diphosphooligosaccharide-protein glycotransferase; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase subunit 48
Gene Aliases: AGER1; CDG1R; DDOST; KIAA0115; OK/SW-cl.45; OKSWcl45; OST; OST48; WBP1
UniProt ID: (Human) P39656
Entrez Gene ID: (Human) 1650
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.