Invitrogen
Manufacturer Code:PA524725
Catalog # PIPA524725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 102-130 amino acids from the central region of human DDO |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aspartic oxidase; D-aspartate oxidase; D-aspartate oxidase, DDO; DASOX; DASOX DDO-1 DDO-2 D-aspartate oxidase DDO aspartic oxidase
Gene Aliases: 5330420D20Rik; 5730402C02Rik; AI467244; DASOX; DDO; DDO-1; DDO-2
UniProt ID: (Human) Q99489, (Mouse) Q922Z0
Entrez Gene ID: (Human) 8528, (Mouse) 70503
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.