Invitrogen
Manufacturer Code:PA513252
Catalog # PIPA513252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 20-49 amino acids from the N-terminal region of human DDIT4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; HIF-1 responsive protein RTP801; Protein regulated in development and DNA damage response 1; Protein regulated in development and DNA damage response 1 REDD-1 HIF-1 responsive protein RTP801; REDD-1
Gene Aliases: DDIT4; Dig2; REDD-1; REDD1; RTP801
UniProt ID: (Human) Q9NX09
Entrez Gene ID: (Human) 54541
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.