Invitrogen
Manufacturer Code:PA554705
Catalog # PIPA554705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DDHD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDHD domain-containing protein 2; intracellular phospholipase A1gamma; KIAA0725p; PA-PLA1 like; Phospholipase DDHD2; SAM, WWE and DDHD domain-containing protein 1; SAMWD1 DDHD domain-containing protein 2 PA-PLA1 like SAM WWE and DDHD domain containing 1 SAM WWE and DDHD domain-containing protein 1 phospholipase DDHD2 sec23p-interacting protein p125-like phosphatidic acid-preferring phospholipase A1; sec23p-interacting protein p125-like phosphatidic acid-preferring phospholipase A1
Gene Aliases: DDHD2; iPLA(1)gamma; KIAA0725; SAMWD1; SPG54
UniProt ID: (Human) O94830
Entrez Gene ID: (Human) 23259
Molecular Function: hydrolase lipase membrane traffic protein phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.