Proteintech
Manufacturer Code:252031AP150UL
Catalog # 50-173-4784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DDHD2 Fusion Protein Ag18373 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDHD domain containing 2; DDHD domain containing 2 DDHD2 KIAA0725 KIAA0725p Phospholipase DDHD2 SAMWD1; DDHD domain-containing protein 2; intracellular phospholipase A1gamma; KIAA0725p; PA-PLA1 like; Phospholipase DDHD2; SAM, WWE and DDHD domain-containing protein 1; sec23p-interacting protein p125-like phosphatidic acid-preferring phospholipase A1
Gene Aliases: 2010305K11Rik; DDHD2; iPLA(1)gamma; KIAA0725; mKIAA0725; SAMWD1; SPG54
UniProt ID: (Human) O94830, (Mouse) Q80Y98
Entrez Gene ID: (Human) 23259, (Mouse) 72108, (Rat) 680971
Molecular Function:
hydrolase
lipase
membrane traffic protein
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.