Novus Biologicals
Manufacturer Code:NBP275465
Catalog # NBP275465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JU32-35 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: damage-specific DNA binding protein 1 (127kD) damage-specific DNA binding protein 1 127kDa Damage-specific DNA-binding protein 1 DDB p127 subunit DDBa DNA damage-binding protein 1 DNA damage-binding protein a HBV X-associated protein 1 UV-damaged DNA-binding factor UV-damaged DNA-binding protein 1 UV-DDB 1 UV-DDB1 XAP1 XAP-1 Xeroderma pigmentosum group E-complementing protein XPCe XPE XPE-BF XPE-binding factor; damage-specific DNA binding protein 1, 127kDa; Damage-specific DNA-binding protein 1; DDB p127 subunit; DDBa; DNA damage-binding protein 1; DNA damage-binding protein a; HBV X-associated protein 1; UV-damaged DNA-binding factor; UV-damaged DNA-binding protein 1; UV-DDB 1; XAP-1; Xeroderma pigmentosum group E-complementing protein; XPCe; XPE-BF; XPE-binding factor
Gene Aliases: DDB1; DDBA; UV-DDB1; XAP1; XPCE; XPE; XPE-BF
UniProt ID: (Human) Q16531
Entrez Gene ID: (Human) 1642
Molecular Function:
DNA binding protein
RNA binding protein
damaged DNA-binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.