OriGene
Manufacturer Code:TA890041
Catalog # 50-168-4754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human DDB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: damage-specific DNA binding protein 1, 127kDa; Damage-specific DNA-binding protein 1; damage-specific DNA-binding protein, DNA repair; damaged-DNA recognition protein 1; DDB p127 subunit; DDBa; DDBA UV-DDB1 XAP1 XPCE XPE XPE-BF; DNA damage-binding protein 1; DNA damage-binding protein a; DNA repair protein; HBV X-associated protein 1; UV-damaged DNA-binding factor; UV-damaged DNA-binding protein 1; UV-DDB 1; XAP-1; Xeroderma pigmentosum group E-complementing protein; XPCe; XPE-BF; XPE-binding factor
Gene Aliases: 127kDa; AA408517; DDB1; DDBA; p127-Ddb1; UV-DDB1; XAP1; XPCE; XPE; XPE-BF
UniProt ID: (Human) Q16531, (Mouse) Q3U1J4
Entrez Gene ID: (Human) 1642, (Mouse) 13194
Molecular Function:
DNA binding protein
RNA binding protein
damaged DNA-binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.