Novus Biologicals
Manufacturer Code:NBP271906JF646
Catalog # N271906J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7D11 |
Immunogen | Full length recombinant protein of human DCXR (NP_057370) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: carbonyl reductase 2; Carbonyl reductase II; Carbonyl reductase II DCR dicarbonyl/L-xylulose reductaseP34H EC 1.1.1 EC 1.1.1.10 HCR2 HCRII human carbonyl reductase 2 KIDCR Kidney dicarbonyl reductase L-xylulose reductase SDR20C1 short chain dehydrogenase/reductase family 20C member 1 Sperm surface protein P34H XR; Dicarbonyl/L-xylulose reductase; kiDCR; Kidney dicarbonyl reductase; L-xylulose reductase; Short chain dehydrogenase/reductase family 20C member 1; short chain dehydrogenase/reductase family 20C, member 1; Sperm surface protein P34H; XR
Gene Aliases: DCR; DCXR; HCR2; HCRII; KIDCR; P34H; PNTSU; SDR20C1; XR
UniProt ID: (Human) Q7Z4W1
Entrez Gene ID: (Human) 51181
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.