Invitrogen
Manufacturer Code:PA570886
Catalog # PIPA570886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse DCUN1D2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13orf17 DCN1-like protein 2 DCUN1 domain-containing protein 2 defective in cullin neddylation protein 1-like protein 2; DCN1, defective in cullin neddylation 1, domain containing 2; DCN1-like protein 2; DCNL2; DCUN1 domain-containing protein 2; Defective in cullin neddylation protein 1-like protein 2; squamous cell carcinoma-related oncogene
Gene Aliases: AW495713; C13orf17; DCUN1D2; DCUN1L2; mFLJ10704; RGD1311743
UniProt ID: (Human) Q6PH85, (Mouse) Q8BZJ7
Entrez Gene ID: (Human) 55208, (Horse) 100067313, (Dog) 476997, (Rabbit) 100355008, (Guinea pig) 100726054, (Rat) 688913, (Mouse) 102323
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.