Novus Biologicals
Manufacturer Code:NBP297034AF750
Catalog # NB041690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant DCUN1D2 (NP_001014305.1 Met1-Phe259). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13orf17 chromosome 13 open reading frame 17 DCN1 defective in cullin neddylation 1 domain containing 2 (S. cerevisiae) DCN1-like protein 2 DCUN1 domain-containing protein 2 DCUN1L2 Defective in cullin neddylation protein 1-like protein 2 FLJ10704 FLJ20092; DCN1, defective in cullin neddylation 1, domain containing 2; DCN1-like protein 2; DCNL2; DCUN1 domain-containing protein 2; Defective in cullin neddylation protein 1-like protein 2
Gene Aliases: C13orf17; DCUN1D2; DCUN1L2
UniProt ID: (Human) Q6PH85
Entrez Gene ID: (Human) 55208
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.