Proteintech
Manufacturer Code:109651AP150UL
Catalog # 50-172-6749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DCUN1D1 Fusion Protein Ag1440 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCN1 like protein 1 DCUN1D1 DCUN1L1 RP42 SCCRO SCRO Tes3; DCN1, defective in cullin neddylation 1, domain containing 1; DCN1-like protein 1; DCNL1; DCUN1 domain-containing protein 1; Defective in cullin neddylation protein 1-like protein 1; RP42 homolog; squamous cell carcinoma related oncogene; Squamous cell carcinoma-related oncogene; testis derived transcript 3; Testis-specific protein 3
Gene Aliases: DCN1; DCNL1; DCUN1D1; DCUN1L1; pTes3; RP42; SCCRO; SCRO; Tes3
UniProt ID: (Human) Q8TEX7, (Mouse) Q9QZ73
Entrez Gene ID: (Human) 54165, (Rat) 310324, (Mouse) 114893
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.