Invitrogen
Manufacturer Code:PA5106404
Catalog # PIPA5106404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DCTN6 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU044699 Dctn6 dynactin 6 dynactin subunit 6 Dynactin subunit p27 novel RGD-containing protein p27 p27 dynactin subunit protein WS-3 RGD containing protein WS3 WS-3; dynactin 6; Dynactin subunit 6; Dynactin subunit p27; novel RGD-containing protein; Protein WS-3
Gene Aliases: DCTN6; p27; WS-3; WS3
UniProt ID: (Human) O00399
Entrez Gene ID: (Human) 10671, (Rat) 290798
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.