Invitrogen
Manufacturer Code:PA572571
Catalog # PIPA572571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 62-89 amino acids from the N-terminal region of human DCT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCT; dopachrome delta-isomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; TRP-2; TRP-2 TYRP2 DT L-dopachrome Delta-isomerase L-dopachrome tautomerase TRP2 dopachrome delta-isomerase tyrosinase-related protein 2; TRP2; tyrosinase related protein-2; Tyrosinase-related protein 2; tyrosine-related protein 2
Gene Aliases: DCT; TRP-2; TYRP2
UniProt ID: (Human) P40126
Entrez Gene ID: (Human) 1638
Molecular Function:
isomerase
oxidase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.