Invitrogen
Manufacturer Code:PA552736
Catalog # PIPA552736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DCT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCT; dopachrome delta-isomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; TRP-2; TRP-2 TYRP2 DT L-dopachrome Delta-isomerase L-dopachrome tautomerase TRP2 dopachrome delta-isomerase tyrosinase-related protein 2; TRP2; tyrosinase related protein-2; Tyrosinase-related protein 2; tyrosine-related protein 2
Gene Aliases: DCT; TRP-2; TYRP2
UniProt ID: (Human) P40126
Entrez Gene ID: (Human) 1638
Molecular Function:
isomerase
oxidase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.