Invitrogen
Manufacturer Code:PA5101647
Catalog # PIPA5101647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TRP2/DCT corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCT; DCT DOPAchrome conversion factor dopachrome delta-isomerase DOPAchrome isomerase DOPAchrome oxidoreductase dopachrome tautomerase dopachrome tautomerase (dopachrome delta-isomerase tyrosine-related protein 2) DT hypothetical protein LOC664745 L-dopachrome Delta-isomerase L-dopachrome isomerase L-dopachrome tautomerase melanogenic enzyme tyrosinase-related protein 2 trp2 RGD1564975 slaty SLATY locus protein slt trp2 TRP-2 tyrosinase related protein-2 tyrosinase-related protein 2 tyrosinase-related protein-2 tyrosine-related protein 2 Tyrp2 Tyrp-2 zgc:109860; DOPAchrome conversion factor; dopachrome delta-isomerase; DOPAchrome isomerase; DOPAchrome oxidoreductase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; SLATY locus protein; TRP-2; TRP2; tyrosinase related protein-2; Tyrosinase-related protein 2; tyrosinase-related protein-2; tyrosine-related protein 2
Gene Aliases: DCT; DT; slaty; slt; TRP-2; TRP2; Tyrp-2; TYRP2
UniProt ID: (Human) P40126, (Mouse) P29812
Entrez Gene ID: (Human) 1638, (Mouse) 13190
Molecular Function:
isomerase
oxidase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.