OriGene
Manufacturer Code:TA505626
Catalog # 50-167-6071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H8 |
Immunogen | Full length human recombit protein of human DCPS produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; ARS DCS1 HINT-5 HINT5 HSL1 HSPC015; DCS-1; Decapping scavenger enzyme; heat shock-like protein 1; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; Histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; m7GpppX diphosphatase; mRNA decapping enzyme; Scavenger mRNA-decapping enzyme DcpS
Gene Aliases: ARS; DCPS; DCS1; HINT-5; HINT5; HSL1; HSPC015
UniProt ID: (Human) Q96C86
Entrez Gene ID: (Human) 28960
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.