Invitrogen
Manufacturer Code:PA5115102
Catalog # PIPA5115102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Dcp2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410015D23Rik 5730537H01Rik AL118268 Dcp2 DCP2 decapping enzyme homolog DCP2 decapping enzyme homolog (S. cerevisiae) decapping mRNA 2 Decapping protein 2 hDpc m7GpppN-mRNA hydrolase mRNA-decapping enzyme 2 Nucleoside diphosphate-linked moiety X motif 20 nudix (nucleoside diphosphate linked moiety X)-type motif 20 Nudix motif 20 nudix-type motif 20 NUDT20 RGD1562909; DCP2 decapping enzyme homolog; hDpc; m7GpppN-mRNA hydrolase; mRNA-decapping enzyme 2; Nucleoside diphosphate-linked moiety X motif 20; nudix (nucleoside diphosphate linked moiety X)-type motif 20; Nudix motif 20
Gene Aliases: 2410015D23Rik; 5730537H01Rik; AL118268; DCP2; NUDT20; RGD1562909
UniProt ID: (Human) Q8IU60, (Mouse) Q9CYC6
Entrez Gene ID: (Human) 167227, (Rat) 291604, (Mouse) 70640
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.