Novus Biologicals
Manufacturer Code:NBP17942220UL
Catalog # NBP17942220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human DCP1BThe immunogen for this antibody is DCP1B. Peptide sequence TLDPEPQHLSLTALFGKQDKATCQETVEPPQTLHQQQQQQQQQQEKLPIR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCP1 DCP1 decapping enzyme homolog B (S. cerevisiae) decapping enzyme hDcp1b EC 3.- FLJ31638 hDcp1b mRNA-decapping enzyme 1B; DCP1 decapping enzyme homolog B; decapping enzyme hDcp1b; mRNA-decapping enzyme 1B
Gene Aliases: DCP1; DCP1B
UniProt ID: (Human) Q8IZD4
Entrez Gene ID: (Human) 196513
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.