Proteintech
Manufacturer Code:138631AP150UL
Catalog # 50-172-9051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DCP1B Fusion Protein Ag4845 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCP1 DCP1B hDcp1b mRNA decapping enzyme 1B; DCP1 decapping enzyme homolog B; decapping enzyme hDcp1b; decapping mRNA 1B; mRNA-decapping enzyme 1B
Gene Aliases: B930050E02Rik; DCP1; DCP1B; RGD1562214
UniProt ID: (Human) Q8IZD4
Entrez Gene ID: (Human) 196513, (Rat) 500305, (Mouse) 319618
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.