Proteintech
Manufacturer Code:223731AP150UL
Catalog # 50-173-3851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DCP1A Fusion Protein Ag18027 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCP1 decapping enzyme homolog A; DCP1 decapping enzyme-like protein A; DCP1A HSA275986 mRNA decapping enzyme 1A Nbla00360 SMAD4IP1 SMIF Transcription factor SMIF; decapping enzyme; decapping enzyme hDcp1a; MAD homolog 4 interacting transcription coactivator 1; MAD homolog 4-interacting transcription coactivator 1; mRNA-decapping enzyme 1A; putative protein product of Nbla00360; Smad4-interacting transcriptional co-activator; Transcription factor SMIF
Gene Aliases: 1110066A22Rik; 4930568L04Rik; AU019772; D14Ertd817e; DCP1A; HSA275986; Mitc1; Nbla00360; SMAD4IP1; SMIF
UniProt ID: (Human) Q9NPI6, (Mouse) Q91YD3
Entrez Gene ID: (Human) 55802, (Mouse) 75901
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.