Invitrogen
Manufacturer Code:MA525400
Catalog # PIMA525400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-280 of DCLRE1B produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5' exonuclease Apollo; APOLLO SNM1B SNMIB 5&prime exonuclease Apollo DNA cross-link repair 1B (PSO2 homolog S. cerevisiae) PSO2 homolog SNM1 homolog B; DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1B protein; PSO2 homolog; SNM1 homolog B; SNMIB
Gene Aliases: APOLLO; DCLRE1B; SNM1B; SNMIB
UniProt ID: (Human) Q9H816
Entrez Gene ID: (Human) 64858
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.