Novus Biologicals
Manufacturer Code:NBP272152JF549
Catalog # N272152J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B7 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-280 of human DCLRE1B (NP_073747) produced in E.coli. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 5' exonuclease Apollo; APOLLO DKFZp686C24187 DNA cross-link repair 1B DNA cross-link repair 1B (PSO2 homolog S. cerevisiae) DNA cross-link repair 1B protein EC 3.1 FLJ12810 FLJ13998 hSNM1B PSO2 homolog SNM1 homolog B SNM1B5&prime exonuclease Apollo SNMIB; DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1B protein; PSO2 homolog; SNM1 homolog B; SNMIB
Gene Aliases: APOLLO; DCLRE1B; SNM1B; SNMIB
UniProt ID: (Human) Q9H816
Entrez Gene ID: (Human) 64858
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.