Proteintech
Manufacturer Code:132031AP150UL
Catalog # 50-172-8567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DCLRE1B Fusion Protein Ag3945 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5 exonuclease Apollo APOLLO DCLRE1B hSNM1B SNM1 homolog B SNM1B SNMIB; 5' exonuclease Apollo; DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1B protein; PSO2 homolog; SNM1 homolog B; SNMIB
Gene Aliases: AI452214; APOLLO; DCLRE1B; mSNM1B; SNM1B; SNMIB
UniProt ID: (Human) Q9H816, (Rat) Q4KLY6, (Mouse) Q8C7W7
Entrez Gene ID: (Human) 64858, (Rat) 310745, (Mouse) 140917
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.