Invitrogen
Manufacturer Code:PA571370
Catalog # PIPA571370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human DCLRE1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA cross-link repair 1A protein; DNA-crosslink repair gene SNM1; hSNM1; PSO2 SNM1 SNM1A DNA cross-link repair 1A (PSO2 homolog S. cerevisiae) DNA cross-link repair 1A protein DNA-crosslink repair gene SNM1 PSO2 homolog SNM1 homolog A hSNM1A; SNM1 homolog A
Gene Aliases: DCLRE1A; KIAA0086; PSO2; SNM1; SNM1A
UniProt ID: (Human) Q6PJP8
Entrez Gene ID: (Human) 9937
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.