Invitrogen
Manufacturer Code:PA557868
Catalog # PIPA557868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DCLRE1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA cross-link repair 1A protein; DNA-crosslink repair gene SNM1; hSNM1; PSO2 SNM1 SNM1A DNA cross-link repair 1A (PSO2 homolog S. cerevisiae) DNA cross-link repair 1A protein DNA-crosslink repair gene SNM1 PSO2 homolog SNM1 homolog A hSNM1A; SNM1 homolog A
Gene Aliases: DCLRE1A; KIAA0086; PSO2; SNM1; SNM1A
UniProt ID: (Human) Q6PJP8
Entrez Gene ID: (Human) 9937
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.