Invitrogen
Manufacturer Code:PA513452
Catalog # PIPA513452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 143-170 amino acids from the central region of human DCL-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin BIMLEC; C-type lectin domain family 13 member A; C-type lectin domain family 13, member A; CD302; CD302 antigen; CD302 antigen C-type lectin domain family 13 member A C-type lectin BIMLEC Type I transmembrane C-type lectin receptor DCL-1 CD302 KIAA0022 DCL1 CLEC13A; DEC205-associated C-type lectin 1; Type I transmembrane C-type lectin receptor DCL-1
Gene Aliases: 1110055L24Rik; AI159627; BIMLEC; CD302; CLEC13A; DCL-1; DCL1; KIAA0022
UniProt ID: (Human) Q8IX05, (Mouse) Q9DCG2
Entrez Gene ID: (Human) 9936, (Mouse) 66205
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.