Novus Biologicals
Manufacturer Code:NBP216107
Catalog # NBP216107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human DCI. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3,2 trans-enoyl-Coenzyme A isomerase; 3,2-trans-enoyl-CoA isomerase; 3,2-trans-enoyl-CoA isomerase, mitochondrial; acetylene-allene isomerase; acetylene-allene isomerase D2-enoyl-CoA isomerase D3 DCI32-trans-enoyl-CoA isomerase32 trans-enoyl-CoA isomerase Delta(2)-enoyl-CoA isomerase Delta(3) delta3 delta2-enoyl-CoA isomerase dodecenoyl-CoA delta isomerase (32 trans-enoyl-CoA isomerase) dodecenoyl-CoA isomerase32-trans-enoyl-CoA isomerase mitochondrial dodecenoyl-Coenzyme A delta isomerase (32 trans-enoyl-Coenzyme A isomerase) EC 5.3.3.8 enoyl-CoA delta isomerase 132 trans-enoyl-Coenzyme A isomerase; D3,D2-enoyl-CoA isomerase; Delta(3),Delta(2)-enoyl-CoA isomerase; dodecenoyl-CoA delta isomerase (3,2 trans-enoyl-CoA isomerase); Dodecenoyl-CoA isomerase; dodecenoyl-Coenzyme A delta isomerase (3,2 trans-enoyl-Coenzyme A isomerase); Enoyl-CoA delta isomerase 1, mitochondrial
Gene Aliases: DCI; ECI1
UniProt ID: (Human) P42126
Entrez Gene ID: (Human) 1632
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.