Invitrogen
Manufacturer Code:PA562158
Catalog # PIPA562158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DCHS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cadherin-19; Cadherin-25; cadherin-related family member 6; fibroblast cadherin 1; fibroblast cadherin FIB1; Fibroblast cadherin-1; Protein dachsous homolog 1; Protocadherin-16
Gene Aliases: CDH19; CDH25; CDHR6; DCHS1; FIB1; KIAA1773; MVP2; PCDH16; VMLDS1
UniProt ID: (Human) Q96JQ0
Entrez Gene ID: (Human) 8642
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.