OriGene
Manufacturer Code:TA804517
Catalog # 50-168-0281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D10 |
Immunogen | Full length human recombit protein of human DCC produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Colorectal cancer suppressor; colorectal tumor suppressor; CRC18 CRCR1 HGPPS2 IGDCC1 MRMV1 NTN1R1; deleted in colorectal cancer protein; deleted in colorectal carcinoma; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; Netrin receptor DCC; putative colorectal tumor suppressor; Tumor suppressor protein DCC
Gene Aliases: CRC18; CRCR1; DCC; IGDCC1; MRMV1; NTN1R1
UniProt ID: (Human) P43146
Entrez Gene ID: (Human) 1630
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.