R&D Systems
Manufacturer Code:MAB5884SP
Catalog # MAB5884SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 531505 |
Immunogen | <EM>E. coli</EM>-derived recombinant human DCC Ser1323-Phe1447 Accession # P43146 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after opening. 6 months -20 to -70 degreesC under sterile conditions |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Colorectal cancer suppressor; Colorectal cancer suppressor CRC18 CRCR1 deleted in colorectal cancer protein deleted in colorectal carcinoma IGDCC1colorectal tumor suppressor Immunoglobulin superfamily DCC subclass member 1 immunoglobulin superfamily DCC subclass member 1 netrin receptor DCC Tumor suppressor protein DCC; colorectal tumor suppressor; deleted in colorectal cancer protein; deleted in colorectal carcinoma; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; Netrin receptor DCC; putative colorectal tumor suppressor; Tumor suppressor protein DCC
Gene Aliases: CRC18; CRCR1; DCC; IGDCC1; MRMV1; NTN1R1
UniProt ID: (Human) P43146
Entrez Gene ID: (Human) 1630
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.