Invitrogen
Manufacturer Code:PA579139
Catalog # PIPA579139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human DCC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Colorectal cancer suppressor; colorectal tumor suppressor; deleted in colorectal cancer protein; deleted in colorectal carcinoma; Immunoglobulin superfamily DCC subclass member 1; immunoglobulin superfamily, DCC subclass, member 1; Netrin receptor DCC; Netrin receptor DCC Colorectal cancer suppressor Immunoglobulin superfamily DCC subclass member 1 Tumor suppressor protein DCC DCC IGDCC1; putative colorectal tumor suppressor; Tumor suppressor protein DCC
Gene Aliases: C030036D22Rik; CRC18; CRCR1; DCC; IGDCC1; MRMV1; NTN1R1
UniProt ID: (Human) P43146, (Mouse) P70211, (Rat) Q63155
Entrez Gene ID: (Human) 1630, (Mouse) 13176, (Rat) 25311
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.