Invitrogen
Manufacturer Code:PA528547
Catalog # PIPA528547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 489 and 750 of Human DCBLD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700055P21Rik; CLCP1 ESDN 1700055P21Rik CUB LCCL and coagulation factor V/VIII-homology domains protein 1 coagulation factor V/VIII-homology domains protein 1 discoidin CUB and LCCL domain-containing protein 2 endothelial and smooth muscle cell-derived neuropilin-like protein; coagulation factor V/VIII-homology domains protein 1; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1; Discoidin, CUB and LCCL domain-containing protein 2; Endothelial and smooth muscle cell-derived neuropilin-like protein
Gene Aliases: CLCP1; DCBLD2; ESDN
UniProt ID: (Human) Q96PD2
Entrez Gene ID: (Human) 131566
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.