Invitrogen
Manufacturer Code:PA520910
Catalog # PIPA520910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the carboxy terminus of human DCLK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CaMK-like CREB regulatory kinase 2; CL2; CL2 CLICK-II CLICK2 CLIK2 DCAMKL2 DCDC3 DCDC3B DCK2 CaMK-like CREB regulatory kinase 2 doublecortin and CaM kinase-like 2 doublecortin domain-containing protein 3B doublecortin-like and CAM kinase-like 2 serine/threonine-protein kinase DCLK2; doublecortin and CaM kinase-like 2; Doublecortin domain-containing protein 3B; doublecortin kinase 2; Doublecortin-like and CAM kinase-like 2; Doublecortin-like kinase 2; Serine/threonine-protein kinase DCLK2
Gene Aliases: 6330415M09Rik; AU044875; CL2; CLICK-II; CLICK2; CLIK2; DCAMKL2; DCDC3; DCDC3B; DCK2; DCLK2; RGD1308384
UniProt ID: (Human) Q8N568, (Rat) Q5MPA9, (Mouse) Q6PGN3
Entrez Gene ID: (Human) 166614, (Rat) 310698, (Mouse) 70762
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.