Novus Biologicals
Manufacturer Code:NBP177128
Catalog # NBP177128
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human DCLK2. The immunogen is located within the last 50 amino acids of DCLK2. Amino Acid Squence: PGGERAGTWRRHRD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CaMK-like CREB regulatory kinase 2; CL2; CL2EC 2.7.11.1 CLICK-II CLIK2 DCDC3 DCDC3BCaMK-like CREB regulatory kinase 2 DCK2DCAMKL2 DKFZp761I032 doublecortin and CaM kinase-like 2 Doublecortin domain-containing protein 3B Doublecortin-like and CAM kinase-like 2 doublecortin-like kinase 2CLICK2 EC 2.7.11 MGC45428 serine/threonine-protein kinase DCLK2; doublecortin and CaM kinase-like 2; Doublecortin domain-containing protein 3B; Doublecortin-like and CAM kinase-like 2; Doublecortin-like kinase 2; Serine/threonine-protein kinase DCLK2
Gene Aliases: CL2; CLICK-II; CLICK2; CLIK2; DCAMKL2; DCDC3; DCDC3B; DCK2; DCLK2
UniProt ID: (Human) Q8N568
Entrez Gene ID: (Human) 166614
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.