Invitrogen
Manufacturer Code:MA524563
Catalog # PIMA524563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G8 |
Immunogen | WDR42A (AAH13107 1 a.a. &bsim 273 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DCAF8 H326 WDR42A; DDB1- and CUL4-associated factor 8; WD repeat domain 42A; WD repeat-containing protein 42A
Gene Aliases: DCAF8; GAN2; H326; WDR42A
UniProt ID: (Human) Q5TAQ9
Entrez Gene ID: (Human) 50717
Molecular Function:
G-protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.