Invitrogen
Manufacturer Code:PA5112994
Catalog # PIPA5112994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human DDB1- and CUL4-associated factor 8 protein (1-135AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DCAF8 DDB1 and CUL4 associated factor 8 DDB1- and CUL4-associated factor 8 GAN2 H326 WD repeat domain 42A WD repeat-containing protein 42A WDR42A; DDB1- and CUL4-associated factor 8; WD repeat domain 42A; WD repeat-containing protein 42A
Gene Aliases: AA408877; AA433120; AA792214; AU018973; C76055; D1Dau35e; D1Ucla4; DCAF8; GAN2; H326; WDR42A
UniProt ID: (Human) Q5TAQ9, (Mouse) Q8N7N5
Entrez Gene ID: (Human) 50717, (Mouse) 98193
Molecular Function:
G-protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.