Invitrogen
Manufacturer Code:PA569818
Catalog # PIPA569818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse DCAF7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AN11 homolog; DDB1 and CUL4 associated factor 7 DCAF7 HAN11 WDR68; DDB1- and CUL4-associated factor 7; dirty south; dys; human anthocyanin; seven-WD-repeat protein of the AN11 family-1; WD repeat domain 68; WD repeat-containing protein 68; WD repeat-containing protein An11 homolog; WD-repeat protein
Gene Aliases: 1700012F10Rik; 2610037L01Rik; AN11; C86529; DCAF7; fj83g06; HAN11; han11l; RGD1305140; SWAN-1; WDR68; wu:fj83g06; zgc:63940
UniProt ID: (Human) P61962, (Mouse) P61963
Entrez Gene ID: (Bovine) 516678, (Horse) 100054273, (Rabbit) 100347842, (Human) 10238, (Dog) 100856670, (Mouse) 71833, (Guinea pig) 100715499, (Rat) 303602, (Zebrafish) 337565
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.