Novus Biologicals
Manufacturer Code:NBP292192100UL
Catalog # NB026275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human DCAF4 (NP_056419.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDB1 and CUL4 associated factor 4 DDB1- and CUL4-associated factor 4 DKFZp434K114 MGC20547 MGC46524 WD repeat domain 21 WD repeat domain 21A WD repeat-containing protein 21A WDR21 WDR21A; DDB1- and CUL4-associated factor 4; WD repeat domain 21A; WD repeat-containing protein 21A
Gene Aliases: DCAF4; WDR21; WDR21A
UniProt ID: (Human) Q8WV16
Entrez Gene ID: (Human) 26094
Molecular Function: RNA binding protein enzyme modulator esterase hydrolase kinase inhibitor kinase modulator mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.