Novus Biologicals
Manufacturer Code:NBP287252
Catalog # NB034874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human DCAF12L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDB1 and CUL4 associated factor 12-like 2; DDB1 And CUL4 Associated Factor 12-Like 2 DDB1- And CUL4-Associated Factor 12-Like Protein 2 WD Repeat Domain 40C WD Repeat-Containing Protein 40C WDR40C; DDB1- and CUL4-associated factor 12-like protein 2; WD repeat-containing protein 40C
Gene Aliases: DCAF12L2; WDR40C
UniProt ID: (Human) Q5VW00
Entrez Gene ID: (Human) 340578
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.