Invitrogen
Manufacturer Code:PA5113943
Catalog # PIPA5113943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human WDR40A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500001L20Rik 5830424K06Rik AA420338 AI851081 cancer/testis antigen 102 Centrosome-related protein TCC52 CT102 Dcaf12 DDB1 and CUL4 associated factor 12 DDB1- and CUL4-associated factor 12 Kiaa1892 LOC682812 similar to WD repeat domain 40A TCC52 Testis cancer centrosome-related protein WD repeat domain 40A WD repeat-containing protein 40A Wdr40a; cancer/testis antigen 102; Centrosome-related protein TCC52; DDB1- and CUL4-associated factor 12; Testis cancer centrosome-related protein; WD repeat domain 40A; WD repeat-containing protein 40A
Gene Aliases: CT102; DCAF12; KIAA1892; TCC52; WDR40A
UniProt ID: (Human) Q5T6F0
Entrez Gene ID: (Human) 25853
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.