Novus Biologicals
Manufacturer Code:NBP299483R
Catalog # NB085340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human DC8. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf48 chromosome 1 open reading frame 48 DC8 DKFZP566O1646 kinetochore-associated protein NSL1 homolog MIS14 NSL1 MIND kinetochore complex component homolog (S. cerevisiae); Kinetochore-associated protein NSL1 homolog; NSL1, MIND kinetochore complex component, homolog
Gene Aliases: C1orf48; DC31; DC8; MIS14; NSL1
UniProt ID: (Human) Q96IY1
Entrez Gene ID: (Human) 25936
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.