Invitrogen
Manufacturer Code:PA524840
Catalog # PIPA524840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DC13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310061C15Rik uncharacterized protein C16orf61; C-x(9)-C motif containing 2; COX assembly mitochondrial protein 2 homolog
Gene Aliases: 1110046L09Rik; 2310061C15Rik; C16orf61; CMC2; DC13
UniProt ID: (Human) Q9NRP2, (Mouse) Q8K199
Entrez Gene ID: (Human) 56942, (Mouse) 66531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.