R&D Systems
Manufacturer Code:FAB7824U100UG
Catalog # FAB7824U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 788524 |
Immunogen | <EM>E. coli</EM>-derived recombinant human DC-STAMP Asp314-Thr376 Accession # Q9H295 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: DC-specific transmembrane protein; DC-STAMP; DCSTAMP dendritic cell-specific transmembrane protein FIND IL-4-induced Protein TM7SF4 transmembrane 7 superfamily member 4; Dendritic cell-specific transmembrane protein; Dendritic cells (DC)-specific transmembrane protein; Dendrocyte-expressed seven transmembrane protein; FIND; IL-4-induced protein; IL-Four (IL-4) induced; IL-Four INDuced; IL-four-induced protein; Transmembrane 7 superfamily member 4
Gene Aliases: DCSTAMP; FIND; hDC-STAMP; TM7SF4
UniProt ID: (Human) Q9H295
Entrez Gene ID: (Human) 81501
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.