R&D Systems
Manufacturer Code:MAB162
Catalog # MAB162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 120604 |
Immunogen | NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR/CD299 Accession # Q9H2X3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; CD209 antigen-like protein 1; CD209L CD209L1 CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2 CD299 CD299 antigenMGC129964 CLEC4M C-type lectin domain family 4 member M DC-SIGN2 DC-SIGN2CD209 antigen-like protein 1 DCSIGNR DC-SIGNRDC-SIGN-related protein DCSIGNRMGC47866 Dendritic cell-specific ICAM-3-grabbing non-integrin 2 HP10347 liver/lymph node-specific ICAM-3 grabbing non-integrin Liver/lymph node-specific ICAM-3-grabbing non-integrin LSIGN L-SIGN LSIGNC-type lectin domain family 4 member M mannose binding C-type lectin DC-SIGNR; CD299; CD299 antigen; DC-SIGN-related protein; DC-SIGN2; DC-SIGNR; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; L-SIGN; liver/lymph node-specific ICAM-3 grabbing non-integrin; Liver/lymph node-specific ICAM-3-grabbing non-integrin; mannose binding C-type lectin DC-SIGNR
Gene Aliases: CD209L; CD209L1; CD299; CLEC4M; DC-SIGN2; DC-SIGNR; DCSIGNR; HP10347; L-SIGN; LSIGN
UniProt ID: (Human) Q9H2X3
Entrez Gene ID: (Human) 10332
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.